VOA Calibration Mix #3 (10 Components), Restek
Catalog # RK30008
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Description:VOA Calibration Mix #3
- Size:1 ml/Ampule
- Cat. no.:RK30008
- Certification:CRM
- Supplier No.:30008
Specifications
About this item
Mix contains: Carbon tetrachloride (56-23-5), Chlorobenzene (108-90-7), Chloroform (67-66-3), 1,1-Dichloroethane (75-34-3), 1,1-Dichloroethene (75-35-4), 1,2-Dichloropropane (78-87-5), Methylene chloride (dichloromethane) (75-09-2), 1,1,2-Trichloroethane (79-00-5), Trichloroethene (79-01-6), m-Xylene (108-38-3).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.