Order Entry
Canada
ContactUsLinkComponent
VOA Calibration Mix #1 (Ketones) (4 Components), Restek
VOA Calibration Mix #1 (Ketones) (4 Components), Restek
Catalog # RK30006
Supplier:  Restek
CAS Number:  
undefined
VOA Calibration Mix #1 (Ketones) (4 Components), Restek
Catalog # RK30006
Supplier:  Restek
Supplier Number:  30006
CAS Number:  

Specifications

  • Concentration:
    5000 µg/ml each in P and T methanol:water (90:10)
  • Description:
    VOA Calibration Mix #1 (ketones)
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK30006
  • Certification:
    CRM
  • Supplier No.:
    30006

Specifications

About this item

Mix contains Acetone (67-64-1), 2-Butanone (MEK) (78-93-3), 2-Hexanone (591-78-6), 4-Methyl-2-pentanone (MIBK) (108-10-1).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.