- Concentration:2000 µg/ml each in methylene chloride
- Description:8270 Calibration Mix #3
- Size:1 ml/Ampule
- Cat. no.:10118-474
- Certification:CRM
- Supplier No.:31620
Contains: Aramite (140-57-8); Bis(2-chloroethyl)ether (111-44-4); Bis(2-chloroethoxy)methane (111-91-1); 4-Bromophenyl phenyl ether (101-55-3); Chlorobenzilate (510-15-6); 2-Chloronaphthalene (91-58-7); 4-Chlorophenyl phenyl ether (7005-72-3); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 1,3-Dinitrobenzene (99-65-0); Hexachlorobenzene (118-74-1); Hexachlorobutadiene (87-68-3); Hexachlorocyclopentadiene (77-47-4); Hexachloroethane (67-72-1); Hexachloropropene (1888-71-7); Isodrin (465-73-6); Kepone (143-50-0); 2,2'-Oxybis(1-chloropropane) (108-60-1); Pentachlorobenzene (608-93-5); Pentachloronitrobenzene (quintozene) (82-68-8); 1,2,4,5-Tetrachlorobenzene (95-94-3); 1,2,4-Trichlorobenzene (120-82-1)