Redox Buffer Solution
Product Details & Documents
Redox buffer solutions are used for verification of all common redox sensors.
- Temperature dependent
- Used in quality assurance systems
A table on every bottle label indicates redox values at different temperatures and hence not used for calibration purposes.
Delivery information: Supplied with a quality inspection certificate.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Description:Redox buffer solution, 220 mV
- Size:250 ml
- Packed:1×250 ml
- Reference temperature:25 °C
- pH:7
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



