Order Entry
Canada
ContactUsLinkComponent
BipPulverizer System II, MP Biomedicals
BipPulverizer System II, MP Biomedicals
  CA76046-292
 :  
undefined
BipPulverizer System II, MP Biomedicals
  CA76046-292
 :  116850200
 :  

 

  • Description:
    BipPulverizer System II
  • Size:
    50×2 ml tubes
  • Storage conditions:
    Store at Room Temperature
  • Cat. no.:
    CA76046-292
  • Application:
    Nucleic Acid Isolation and Purification

 

 

BioPulverizer™ System II contains 10x2 mL tubes of each of our specialized Lysing Matrices.

  • Contains Matrix types F, G, H, I, and J
  • This sampler pack gets you going
  • Optimizes homogenization
  • Ideal for very difficult, unusual, or multiple samples

Contains 10 each of the impact-resistant 2 mL tube of: Lysing Matrix F, Lysing Matrix G, Lysing Matrix H, Lysing Matrix I, and Lysing Matrix J.

Matrix F contains 1.6 mm aluminum oxide granules and 1.6mm silicon carbide granules. Matrix G contains 1.6mm silicon carbide granules and 2 mm glass beads. Matrix H contains 2 mm glass and 2 mm yellow zirconium oxide beads. Matrix I contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead. Matrix J contains 2 mm yellow zirconia beads and 1.6mm aluminum oxide granules.

 : DNase and RNase Free