Order Entry
Canada
ContactUsLinkComponent
Herbicide Mix #2, Derivatized Form, Restek
Herbicide Mix #2, Derivatized Form, Restek
Supplier:  Restek
Certificates
undefined
Herbicide Mix #2, Derivatized Form, Restek
Supplier:  Restek

Specifications

  • CAS No.:
    17640-02-7
  • Formula:
    C₄H₆Cl₂O₂
  • Description:
    Dalapon methyl ester

Specifications

Frequently Bought Together