LB Agar Plates
LB Agar plates, 20 plates, 100 mm
Catalog # CA77443-320
New Product
Supplier: Teknova
Supplier #: L1096
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
1.69% Animal-free soytone, 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar 50 µg/ml kanamycin.
- Shipping conditions: Ambient
- Storage conditions: Refrigerate
For use with kanamycin resistant strains and strains harboring plasmids such as pPIC9K, pCR®-Blunt, pUC9P series plasmids. Used as a substitute for LB Agar when the media cannot contain any animal products.
Specifications
Specifications
- Description:LB Agar plates, 20 plates
- Plate diameter:100 mm
- Cat. No.:CA77443-320
- Sterility:Sterile
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



