Tryptic Soy Agar Plates
Supplier: Teknova
About this item
Tryptic Soy Agar Plates contains 1.5% Tryptone, 0.5% soytone, 0.5% NaCl, and 1.5% agar used for isolating and cultivating a variety of fastidious microorganisms.
2 Options Available Below
Product Details & Documents
Tryptic Soy Agar Plates contains 1.5% Tryptone, 0.5% soytone, 0.5% NaCl, and 1.5% agar used for isolating and cultivating a variety of fastidious microorganisms.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA77442-962
- CA100218-562
- Sterility:
- Sterile
- Sterile
Specifications
Product Family Options
Product Information
- Plate sizeSizeAvailabilityPrice
- CA77442-962Tryptic soy agar platesSupplier Number: T0147Specifications:New Product
More
Specifications
Cat. No.CA77442-962SterilitySterileNew Product - CA100218-562Tryptic soy agar platesSupplier Number: T0147Specifications:
More
Specifications
Cat. No.CA100218-562SterilitySterile