Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant DPYSL2 (from Wheat germ)
 :  Abnova
 
Human Recombinant DPYSL2 (from Wheat germ)
 :  Abnova

 

  • Protein/peptide type:
    Recombinant
  • Source:
    Wheat germ
  • Species:
    Human
  • Storage conditions:
    –80 °C
  • Protein/peptide name:
    DPYSL2
  • Molecular weight:
    88.7
  • Tested applications:
    ELISA, Western Blot, Antibody Production, Protein Array

 

Frequently Bought Together