Specifications
- Material:Stainless Steel
- Cat. no.:89023-022
- Length:63.5 mm (2 1/2")
- Supplier No.:21-060
- Blade Type:Smooth, Sharp/Sharp, Straight
Specifications
About this item
These scissors are straight and have one serrated blade and one smooth blade.
- Reusable
- Small
Econo™ Utility Scissors are multipurpose instruments used for cutting non-sterile materials such as dressings, casts, and surgical drapes. They are Sklar's floor-grade instruments and are sold in bulk quantities. This item is sold in boxes of 12.