MagniTouch Loupe
Catalog #: 470227-882
Supplier: CARSON OPTICAL
Specifications
- Magnification:3×
- Description:MagniTouch
- Cat. no.:470227-882
Specifications
About this item
A 3X touch activated loupe.
- 3 super bright LED's
- Crystal clear glass lens
- On/off switch
- 2 CR2016 batteries
- Twist-action focus
Customers Who Bought This Also Bought