LumiLoupe
Catalog #: 470227-874
Supplier: CARSON OPTICAL
Specifications
- Magnification:17.5×
- Description:LumiLoupe
- Cat. no.:470227-874
Specifications
About this item
A loupe with a transparent base to allow ambient light to illuminate the viewing object.
- Allows for viewing of an object's fine detail
- Dual lenses for close-up viewing and less distortion
Customers Who Bought This Also Bought