BoneClones® Skull Trauma Set of Six Fragments
Catalog # 470228-132
Supplier: BONE CLONES HOLDINGS
Specifications
- Description:Skull Trauma Set of Six Fragments
- Cat. no.:470228-132
Specifications
About this item
A blunt force trauma set comprising six skull fragments from six individuals.
- From The Maxwell Museum of Anthropology's Laboratory of Human Osteology
- Shows depressed fractures at various stages of healing
- Inner and outer tables of the skull are visible
These fragments show various types of depressed fractures at various stages of healing (from no evidence of healing to well-healed fractures). The injuries were caused by different weapons (from hammers to bullets). In addition, the injuries occurred on different parts of the skull; and since the skulls are presented as fragments, one can also see the inner and outer tables of the skulls around the sites of the injuries.
Ordering information: The osteological evaluation report (PDF) will be sent with the purchase of this specimen.
Please note that specimens from the Maxwell Collection can only be sold to recognized educational institutions and professionals associated with educational institutions.
Please note that specimens from the Maxwell Collection can only be sold to recognized educational institutions and professionals associated with educational institutions.