Order Entry
Canada
ContactUsLinkComponent
ELUGENT™ Non-Ionic Detergent, Calbiochem®
ELUGENT™ Non-Ionic Detergent, Calbiochem®
Catalog # CA80058-062
Supplier:  MilliporeSigma
CAS Number:  
ELUGENT™ Non-Ionic Detergent, Calbiochem®
Catalog # CA80058-062
Supplier:  MilliporeSigma
Supplier Number:  324707-100ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    ELUGENT™ Detergent
  • Size:
    100 mL
  • Cat. no.:
    CA80058-062
  • Formulation:
    50% aqueous solution
  • Supplier No.:
    324707-100ML

Specifications

About this item

Non-ionic detergent composed of a mixture of alkyl glucosides

  • HLB number: 13.1.
  • Form: Clear, viscous brown to yellow liquid

It is useful for solubilization and purification of membrane-bound proteins. Relatively low cost alternative to n-octyl-β-D-glucopyranoside and n-octyl-β-D-maltoside in large-scale solubilizations.