Microsampling Vials, 8-425, Sun Sri
Supplier: Thermo Scientific
Jump to:
About this item
Microsampling vials are used to work with small samples, or samples that must be evaporated and reconstituted without having to use multiple sample vials.
3 Options Available Below
Product Details & Documents
Microsampling vials are used to work with small samples, or samples that must be evaporated and reconstituted without having to use multiple sample vials.
Product Family Options
Product Information
- CapacityBottom styleDiameter ×HMaterialColorCap includedCap SizeAvailabilityPrice
- 14232-380Microsampling standard screw thread vialsMicrosampling standard screw thread vials0.30 mlConical bottom12×32 mmPPNo8-425Log in for availabilityLog in for availabilityCase of 1,000Case of 1,000 - $1,065.85$1,065.85New Customer Promotional PriceCase of 1,000$1,065.85Supplier Number: 500 114
- 14234-634Microsampling standard screw thread vialsMicrosampling standard screw thread vials0.30 mlConical12×32 mmPPNo8-425Discontinued-Supplier Number: -
- 66009-772Microsampling standard screw thread vialsMicrosampling standard screw thread vials0.25 mlMandrel12×32 mmPPNo8-425Log in for availabilityLog in for availabilityCase of 1,000Case of 1,000 - $1,039.24$1,039.24New Customer Promotional PriceCase of 1,000$1,039.24Supplier Number: 200 264
- 14235-452Microsampling standard screw thread vialsMicrosampling standard screw thread vials0.50 mlTapered12×32 mmPPNo8-425Log in for availabilityCase of 1,000$950.37Supplier Number: 500 120
- 66106-198Microsampling standard screw thread vialsMicrosampling standard screw thread vials0.50 mlTapered12×32 mmPPNo8-425Discontinued-Supplier Number: -
- 14232-788Microsampling standard screw thread vialsMicrosampling standard screw thread vials1.10 mlTapered bottom12×32 mmGlassClearNo8-425Discontinued-Supplier Number: -