Order Entry
Canada
ContactUsLinkComponent
Sucrose (Saccharose) Test Strips, Reflectoquant®, EMD Millipore
Sucrose (Saccharose) Test Strips, Reflectoquant®, EMD Millipore
Catalog # CA1.16141.0001
Supplier:  MilliporeSigma
CAS Number:  
Sucrose (Saccharose) Test Strips, Reflectoquant®, EMD Millipore
Catalog # CA1.16141.0001
Supplier:  MilliporeSigma
Supplier Number:  1.16141.0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Sucrose (Saccharose) Test, reflectometric test strip
  • Cat. no.:
    CA1.16141.0001
  • Range:
    0.25 - 2.50 g/L
  • Supplier No.:
    1.16141.0001
  • No. of Tests:
    50

Specifications

About this item

The Reflectoquant® Sucrose test is suitable for determination of sucrose in beverages, e. g. fruit/vegetable juice and refreshment drinks after appropriate sample pretreatment or dilution.

  • Assay - standard 1: 0.26 - 0.34 mg/L
  • Assay - standard 2: 1.12 - 1.38 mg/L
  • Assay - standard 3: 1.80 - 2.20 mg/L

Sucrose (saccharose) is cleaved by sucrose phosphorylase into fructose and glucose-1-phosphate. The latter is converted by phosphoglucomutase into glucose-6-phosphate. This is in turn oxidized by NAD under the catalytic effect of glucose-6-phosphate dehydrogenase to gluconate-6-phosphate. In the presence of diaphorase the NADH formed in the process reduces a tetrazolium salt to a blue formazan that is determined reflectometrically on the RQflex®. For the determination of only glucose please refer to our MQuant® or Reflectoquant® Glucose tests.

Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.