You searched for: Rotating/Rocking Shakers
Rotating and rocking shakers combine the functions of a laboratory rocker with a shaker. Complete with variable speed settings and a configurable tilt angle, these compact instruments tailor operations for blot washing, gel staining, and hybridization applications. The rotating and rocking units can be placed inside incubators or cold rooms as required. Rotating and rocking shaker options offer single- or double-tiered platforms that are durably constructed and easy-to-clean.
VWR® Tube Rotators
Supplier: VWR International
VWR® Tube rotators are ideal for applications requiring highly efficient and consistent mixing. VWR offers rugged and reliable fixed speed, digital and large capacity models.
Expand 3 Items
VWR® Nutating Mixer
Supplier: VWR International
The three-dimensional shaking motion of this mixer is gentle yet efficient, making it ideal for mixing samples in blood and centrifuge tubes.
Expand 1 Items
VWR® Signature™ 3D Rotator Waver, 230 V
Supplier: VWR International
3-D rotator waver is ideal for blood samples, DNA extractions, blotting techniques, and general mixing of various size tubes.
Expand 1 Items
VWR® 2D Platform Rockers
Supplier: VWR International
VWR® 2D platform rockers are ideal for use in blotting and staining applications as well as gentle mixing of solutions and reagents.
Expand 2 Items
VWR® Variable Speed Rocker
Supplier: VWR International
Designed to provide precise speed and tilt angles for a broad range of molecular and biological mixing applications.
Expand 1 Items
VWR® Tube Rocker Collection
Supplier: VWR International
The VWR® Tube Rocker Collection features Tube Rockers available in a "Mini" size as well as an "XL" size and in Fixed Speed or a Variable Speed models.
Expand 3 Items
VWR® Analog Rocking Platform Shakers
Supplier: VWR International
VWR® Analog Rocking Platform Shakers
Expand 2 Items
VWR® Mini Tube Rotator, 230 V
Supplier: VWR International
The VWR® Mini Tube Rotator provides a wide range of mixing capabilities suitable for an array of laboratory applications.
Expand 1 Items
VWR® Signature™ Rocking Platform Shaker
Supplier: VWR International
Ideal for cell culture work, staining and destaining gels, hybridization procedures, hematology, and blotting techniques.
Expand 2 Items
VWR® Tube Revolvers
Supplier: VWR International
Efficient, powerful tube revolvers fixed or variable speed, with 5 interchangeable rotor series.
Expand 2 Items
VWR® 3D Nutating Mixers
Supplier: VWR International
VWR® 3D nutating mixers are designed for sample mixing with combined motions of an orbital shaker and 2D platform rocker. These mixers are well suited for many routine laboratory applications that require thorough sample mixing.
Expand 4 Items
VWR® Signature™ Rocking Platform Shaker, 230 V
Supplier: VWR International
Ideal for cell culture work, staining and destaining gels, hybridization procedures, hematology, and blotting techniques.
Expand 1 Items
VWR® Signature™ 3D Rotator Waver
Supplier: VWR International
3-D rotator waver is ideal for blood samples, DNA extractions, blotting techniques, and general mixing of various size tubes.
Expand 2 Items
VWR® Mini Tube Rotator
Supplier: VWR International
The VWR® Mini Tube Rotator provides a wide range of mixing capabilities suitable for an array of laboratory applications.
Expand 7 Items
VWR® Analog 3D Waving Platform Shaker, 230 V
Supplier: VWR International
VWR® Analog waving platform shakers are an easy and economical option for all of your waving shaking needs.
Expand 1 Items
VWR® Rotating Mixer XL
Supplier: VWR International
The VWR® rotating mixer XL mixes common laboratory samples gently or vigorously.
Expand 1 Items
VWR® Tube Roller Series
Supplier: VWR International
VWR® tube rollers are designed for gentle yet complete mixing of a wide range of sample vessels and tubes.
Expand 2 Items
VWR® Analog 3D Waving Platform Shaker
Supplier: VWR International
VWR® Analog waving platform shakers are an easy and economical option for all of your waving shaking needs.
Expand 1 Items
VWR® Tube Rotator and Rotisseries
Supplier: VWR International
The unique design of the VWR® Tube Rotator allows for a variety of tube sizes and mixing applications to be performed efficiently on one instrument.
Expand 1 Items
Rotators for Cell Culture, 240 V
Supplier: Ace Glass
This variable speed rotator is driven by a continuous-duty motor that can withstand a demanding schedule and long hours of operation.
Expand 2 Items
Rotators for Cell Culture, Ace Glass
Supplier: Ace Glass
This variable speed rotator is driven by a continuous-duty motor that can withstand a demanding schedule and long hours of operation.
Expand 1 Items
Corning® LSE™ Nutating Mixer, Corning
Supplier: Corning
The LSE Nutating mixer is a small personal sized 3D mixer which is ideal for ambient and cold room applications for example immunohistochemistry, mixing and resupending small containers such as blood and centrifuge tubes.
Expand 1 Items
Corning® LSE™ Platform Rockers, Corning
Supplier: Corning
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications.
Expand 2 Items
CELLSPIN Spinner System, Argos Technologies
Supplier: Argos Technologies
CELLSPIN a reliable cell culture system.
Expand 12 Items
Corning® LSE™ Platform Rockers, 230 V
Supplier: Corning
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications.
Expand 2 Items
Digital Mini Rotators, Thermo Scientific
Supplier: Thermo Fisher Scientific
The Thermo Scientific™ Digital mini rotator has been designed for culturing bacteria and fungi, hybridization assays, and gel staining and de-staining.
Expand 1 Items
Digital Tube Rocker, Thermo Scientific
Supplier: Thermo Fisher Scientific
Digital Tube Rocker is a precision-controlled platform mixer that accommodates double capacity in a compact footprint for smooth rocking and uniform suspension.
Expand 1 Items
Digital Mini Rotators, 230 V
Supplier: Thermo Fisher Scientific
The Thermo Scientific™ Digital mini rotator has been designed for culturing bacteria and fungi, hybridization assays, and gel staining and de-staining.
Expand 1 Items
Digital Waving Rotators, 230 V
Supplier: Thermo Fisher Scientific
Digital Waving Rotator is designed for ELISA, protein synthesis, gel staining and de-staining gels, antibody staining, and other time-sensitive applications.
Expand 1 Items
Digital Platform Rocker, Thermo Scientific
Supplier: Thermo Fisher Scientific
The Digital Platform Rocker's double platform accommodates more samples while providing consistent rocking motion and easy angle adjustments.