Order Entry
Canada
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody (redFluor® 710) [clone: 61D3]
Anti-CD14 Mouse Monoclonal Antibody (redFluor® 710) [clone: 61D3]
Catalog # 76040-288
Supplier:  Tonbo Biosciences
undefined
Anti-CD14 Mouse Monoclonal Antibody (redFluor® 710) [clone: 61D3]
Catalog # 76040-288
Supplier:  Tonbo Biosciences
Supplier Number:  80-0149-T025

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD14
  • Antigen symbol:
    CD14
  • Clonality:
    Monoclonal
  • Clone:
    61D3
  • Conjugation:
    redFluor® 710
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    25 tests
  • Gene ID:
    929
  • Antigen synonyms:
    LPS-R, Mo2
  • Storage temperature:
    2-8°C protected from light
  • Cat. no.:
    76040-288

Specifications

About this item

The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: redFluor® 710
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human