Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 76040-356
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Rabbit
- Size:1 mg
- Epitope:Heavy and light chain
- Cross adsorption:Yes
- Storage temperature:Store at 2-8 °C
- Cat. no.:76040-356
Specifications
About this item
Goat anti-Rabbit IgG (H&L) - Affinity Pure, FITC Conjugate, min x w/ bovine, goat, human, mouse or rat IgG or serum
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit