Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
76040-190
- Antibody type:Secondary
- Antigen name:Immunoglobulin M
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Size:2 mg
- Epitope:Heavy and light chain
- Storage temperature:Store at 2-8 °C
- Cat. no.:76040-190
Goat anti-Mouse IgM (H&L) - Affinity Pure, FITC Conjugate
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse