Order Entry
Canada
ContactUsLinkComponent
NAD Nucleotidase (from E. coli)
NAD Nucleotidase (from E. coli)
Catalog # 102980-456
Supplier:  Adipogen
CAS Number:  
NAD Nucleotidase (from E. coli)
Catalog # 102980-456
Supplier:  Adipogen
Supplier Number:  AG-40T-0110-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    NAD Nucleotidase
  • Enzyme activity:
    With AMP: 856.4 nmol of Ade/min/mg of protein
    With NMN: 110,3 nmol of NR/min/mg of protein
    With NAD: 5.4 nmol of Ade/min per mg of protein | 5.3 nmol of NR/min/mg of protein
  • Purity:
    98%
  • Molecular weight:
    ~63.0 kDa
  • Concentration:
    0.5 mg/ml (Lot specific)
  • Formulation:
    In 5mM KH2PO4, pH 6.8 containing 100 mM TRIS-HCl (pH 8.5) and 10 mM MgCl₂.
  • Cat. no.:
    102980-456
  • Supplier No.:
    AG40T0110C050

Specifications

About this item

NadN (NAD nucleotidase) is a periplasmic enzyme from Haemophilus influenzae, a major pathogen of the respiratory tract in humans that has developed the capability to exploit host NAD(P) for its nicotinamide dinucleotide requirement

  • UniProt link Q4QNY4: NAD Nucleotidase (H. influenza)

NadN plays a central role by degrading NAD into adenosine and NR (nicotinamide riboside), NMN to nicotinamide riboside and AMP to adenosine. It shows a broad substrate specificity, recognizing either mono- or di-nucleotide nicotinamides and different adenosine phosphates with a maximal activity on 5'-adenosine monophosphate. It can be used as an economically alternative to produce NR (nicotinamide riboside). Catalytic activity: NAD=AMP + NMN=Ade+NR+PPi | AMP=Ade+Pi | NMN=NR+Pi