Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Source:E. coli
- Size:50 µg
- Storage conditions:–80 °C
- Form:Liquid
- Shipping conditions:–80 °C, dry ice
- Enzyme name:NAD Synthetase
- Enzyme activity:~0.1 U/mg protein. (Ammonium or glutamine as nitrogen source). One unit is defined as the amount of enzyme that synthesizes 1µmol NAD per min.
- Purity:98%
- Molecular weight:~75.0 kDa
- Concentration:0.5 mg/ml (Lot specific)
- Formulation:In 20 mM TRIS-HCl, pH 7.8.
- Cat. no.:102980-452
- Supplier No.:AG40T0108C050
Specifications
About this item
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways
- UniProt link P0A5L6: NAD Synthetase (M. tuberculosis)
Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It can use both glutamine or ammonia as a nitrogen source. Catalytic activity: ATP + deamido-NAD+ + L-glutamine + H2O = AMP + diphosphate + NAD+ + L-glutamate