Order Entry
Canada
ContactUsLinkComponent
NAD Kinase (from E. coli)
NAD Kinase (from E. coli)
  102980-450
 :  Adipogen
 :  
NAD Kinase (from E. coli)
  102980-450
 :  Adipogen
 :  AG-40T-0107-C050
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    NAD Kinase
  • Enzyme activity:
    ~1.2 U/mg protein. (Substrate: NAD (+ ATP))
    ~3.8 U/mg protein. (Substrate: NAD (+ poly(P))
    One unit is defined as the amount of enzyme that synthesizes 1µmol nicotinamide adenine dinucleotide phosphate (NADP) per min.
  • Purity:
    98%
  • Molecular weight:
    ~33.0 kDa
  • Concentration:
    0.5 mg/ml (Lot specific)
  • Formulation:
    In 10 mM TRIS at pH 8.0, containing 1 mM MgCl₂, 0.5 mM EDTA, 1 mM DTT and 1 mM PMSF.
  • Cat. no.:
    102980-450
  • Supplier No.:
    AG40T0107C050

 

 

NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions

  • UniProt link P0A5S6: NAD Kinase (M. tuberculosis)

NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. NAD kinase from Mycobacterium tuberculosis is not ATP-dependent but has a broader substrate specificity than human NAD kinase. It can economically produce NADP+ by using other nucleoside triphosphates as well as inorganic polyphosphate as a source of phosphorus. Was identified as a high-confidence drug target. Catalytic activity: ATP + NAD+ = ADP + NADP+