Order Entry
Canada
ContactUsLinkComponent
NAD Kinase (from E. coli)
NAD Kinase (from E. coli)
Catalog # 102980-448
Supplier:  Adipogen
CAS Number:  
NAD Kinase (from E. coli)
Catalog # 102980-448
Supplier:  Adipogen
Supplier Number:  AG-40T-0106-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    NAD Kinase
  • Enzyme activity:
    ~2.6 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1µmol nicotinamide adenine dinucleotide phosphate (NADP) per min.
  • Purity:
    98%
  • Molecular weight:
    ~30.0 kDa
  • Concentration:
    Lot specific
  • Formulation:
    In 50 mM TRIS-HCl, pH 7.0 containing 150 mM sodium chloride, 1 mM EDTA and 1 mM dithiothreitol.
  • Cat. no.:
    102980-448
  • Supplier No.:
    AG40T0106C050

Specifications

About this item

NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions

  • UniProt link O31612: NAD Kinase (B. subtilis)

NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. NAD kinase from Bacillus subtilis is not ATP-dependent but has a broader substrate specificity than human NAD kinase. It can economically produce NADP+ by using other nucleoside triphosphates as well as inorganic polyphosphate as a source of phosphorus. Catalytic activity: ATP + NAD+ = ADP + NADP+