Order Entry
Canada
ContactUsLinkComponent
NAPRTase (from E. coli)
NAPRTase (from E. coli)
Catalog # 102980-444
Supplier:  Adipogen
CAS Number:  
NAPRTase (from E. coli)
Catalog # 102980-444
Supplier:  Adipogen
Supplier Number:  AG-40T-0104-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    NAPRTase
  • Enzyme activity:
    ~0.8 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1µmol nicotinic acid mononucleotide (NaMN) per min.
  • Purity:
    98%
  • Molecular weight:
    ~56.0 kDa
  • Concentration:
    Lot specific
  • Formulation:
    in 20 mM TRIS-HCl, pH 7.8 containing 50 mM sodium chloride.
  • Cat. no.:
    102980-444
  • Supplier No.:
    AG40T0104C050

Specifications

About this item

Nicotinate Phosphoribosyltransferase (NAPRTase) is involved in the biological processes of pyridine nucleotide biosynthesis and nicotinate nucleotide salvage and functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN)

  • UniProt link O32090: NAPRTase (B. subtilis)

It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.