Order Entry
Canada
ContactUsLinkComponent
TDO (from Escherichia coli), AdipoGen Life Sciences
TDO (from Escherichia coli), AdipoGen Life Sciences
Catalog # 102980-438
Supplier:  Adipogen
CAS Number:  
TDO (from Escherichia coli), AdipoGen Life Sciences
Catalog # 102980-438
Supplier:  Adipogen
Supplier Number:  AG-40T-0101-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    TDO
  • Enzyme activity:
    Specific activity towards L-Trp: 87.0µmol/hr per mg. TDO catalyzes the oxidative opening of the L-tryptophan indole moiety to yield N-formylkynurenine (NFK).
  • Purity:
    98%
  • Molecular weight:
    ~47.9 kDa
  • Concentration:
    0.5 mg/ml (Lot specific)
  • Formulation:
    In 50 mM sodium phosphate pH 8.0, containing 0.15M sodium chloride, 0.2 M imidazole and 10% glycerol.
  • Cat. no.:
    102980-438
  • Supplier No.:
    AG40T0101C050

Specifications

About this item

Tryptophan 2,3-dioxygenase (TDO) is a heme-containing dioxygenase catalyzing the addition of molecular oxygen across the 2,3-double bond of the indole ring of tryptophan to form N-formylkynurenine (NFK)

  • UniProt link O77457: TDO (Anopheles gambiae)

In Anopheles gambiae, TDO is the only enzyme able to catalyze the first and rate-limiting step in L-Trp catabolism through the kynurenine pathway. Tryptophan is an essential amino acid for protein synthesis and also the precursor for production of a number of neurotransmitters, such as serotonin and melatonin; in mosquitoes, the kynurenine pathway is essential for eye pigmentation. Conceivably, the tryptophan-using pathways should be regulated in a coordinated manner in mosquitoes as well as in other species and TDO activation/inactivation processes could play an essential role in these phenomena.