Order Entry
Canada
ContactUsLinkComponent
3-Hydroxykynurenine Transaminase (from E. coli)
3-Hydroxykynurenine Transaminase (from E. coli)
Catalog # 102980-436
Supplier:  Adipogen
CAS Number:  
3-Hydroxykynurenine Transaminase (from E. coli)
Catalog # 102980-436
Supplier:  Adipogen
Supplier Number:  AG-40T-0100-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping conditions:
    –80 °C, dry ice
  • Enzyme name:
    3-Hydroxykynurenine Transaminase
  • Enzyme activity:
    Specific activity towards D,L-3HK: 33 µmol/min per mg. Primarily catalyses the pyridoxal 5'-phosphate (PLP)-dependent irreversible transamination of 3-hydroxykynurenine (3-HK) to xanthurenic acid (XA).
  • Purity:
    99%
  • Molecular weight:
    ~44.3 kDa
  • Concentration:
    0.5 mg/ml (Lot specific)
  • Formulation:
    In 20mM TRIS-HCl pH 8.0, containing 150mM NaCl and 40µM pyridoxal 5'-phosphate (PLP).
  • Cat. no.:
    102980-436
  • Supplier No.:
    AG40T0100C050

Specifications

About this item

The 3-hydroxykynurenine transaminase (3-HKT) gene plays a vital role in the development of malaria parasites by participating in the synthesis of xanthurenic acid, which is involved in the exflagellation of microgametocytes in the midgut of malaria vector species

  • UniProt link Q7PRG3: 3-HKT (Anopheles gambiae)

The 3-HKT enzyme is involved in the tryptophan metabolism of Anophelines. The gene had been studied in the important global malaria vector, Anopheles gambiae.