About this item
Mitochondrial NMNAT isoform
- UniProt link Q96T66: NMNAT3 (human)
Catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. Can use tiazofurin monophosphate as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD+. For the pyrophosphorolytic activity, mitochondrial NMNAT isoform can use NAD(+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP+, NADPH and NAADP+. Protects against axonal degeneration following injury.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Form:Liquid
- Shipping conditions:–20 °C, blue Ice
- Enzyme name:NMNAT3
- Enzyme activity:>2U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1µmol of NAD+ per min.
- Purity:95%
- Molecular weight:~27 kDa (monomer)
- Concentration:1 mg/ml
- Formulation:In 50 mM TRIS-HCl, pH 8.0, containing 300 mM sodium chloride, 1 mM DTT and 10% glycerol.