Order Entry
Canada
ContactUsLinkComponent
Human NAD Kinase (from E. coli)
Human NAD Kinase (from E. coli)
Catalog # 102980-420
Supplier:  Adipogen
CAS Number:  
Human NAD Kinase (from E. coli)
Catalog # 102980-420
Supplier:  Adipogen
Supplier Number:  AG-40T-0090-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Free
  • Form:
    Liquid
  • Shipping conditions:
    –20 °C, blue Ice
  • Enzyme name:
    NAD Kinase
  • Enzyme activity:
    >2 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1 µmol of NADP per min.
  • Purity:
    95%
  • Molecular weight:
    ~42 kDa (monomer)
  • Concentration:
    1 mg/ml
  • Formulation:
    Ammonium sulfate suspension
  • Cat. no.:
    102980-420
  • Supplier No.:
    AG40T0090C050

Specifications

About this item

NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions

  • UniProt link O95544: NAD kinase (human)

NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.