Order Entry
Canada
ContactUsLinkComponent
Mouse IgG2b Isotype Control
Mouse IgG2b Isotype Control
Catalog # 102978-716
Supplier:  Adipogen
Mouse IgG2b Isotype Control
Catalog # 102978-716
Supplier:  Adipogen
Supplier Number:  AG-35B-0008-C050

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Control
  • Conjugation:
    Unconjugated
  • Host:
    HEK293
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Size:
    50 µg
  • Form:
    Lyophilized.
  • Purity:
    95%
  • Storage buffer:
    Contains PBS.
  • Molecular weight:
    ~30kDa (SDS-PAGE)
  • Storage temperature:
    -20 °C
  • Concentration:
    1mg/ml after reconstitution.
  • Shipping temperature:
    -20 °C, Blue Ice
  • Cat. no.:
    102978-716
  • Supplier No.:
    AG35B0008C050

Specifications

About this item

Fc (mouse) IgG2b Control (rec.) (Prod. No. AG-35B-0008) serves as a negative control in experiments with anti-APRIL (mouse), mAb (rec.) (Blocking) (APRY-1-1) (Prod. No. AG-27B-0001).

  • Endotoxin content: <0.1EU/µg
  • Endotoxin level: Low