Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant Tim-3 (from CHO cells)
Mouse Recombinant Tim-3 (from CHO cells)
Catalog # 102981-686
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant Tim-3 (from CHO cells)
Catalog # 102981-686
Supplier:  Adipogen
Supplier Number:  CHI-MF-111T3-C100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    <0.06EU/µg
  • Biological activity:
    Immobilized recombinant human Galectin-9 at 500ng/ml can bind Tim-3 (mouse):Fc (human) (rec.) with an apparent KD <10nM. Optimal dilutions should be determined by each laboratory for each application.
  • Gene ID:
    AAI06852.1
  • Protein synonyms:
    TIMD3|Hepatitis A Virus Cellular Receptor 2|HAVcr-2|T Cell Immunoglobulin and Mucin Domain-containing Protein 3|TIM3
  • Protein/peptide name:
    Tim-3
  • Purity:
    98%
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping temperature:
    -20 °C, Blue Ice
  • Cat. no.:
    102981-686
  • Supplier No.:
    CHIMF111T3C100

Specifications

About this item

The TIM (T cell/transmembrane, immunoglobulin and mucin) family plays a critical role in regulating immune responses, including allergy, asthma, transplant tolerance, autoimmunity and the response to viral infections. The unique structure of TIM immunoglobulin variable region domains allows highly specific recognition of phosphatidylserine (PtdSer), exposed on the surface of apoptotic cells. Tim-3, a type I transmembrane protein, contains an immunoglobulin and a mucin-like domain in its extracellular portion and a tyrosine phosphorylation motif in its cytoplasmic portion. TIM-3 is preferentially expressed on Th1 and Tc1 cells, and generates an inhibitory signal resulting in apoptosis of Th1 and Tc1 cells. TIM-3 is also expressed on some dendritic cells and can mediate phagocytosis of apoptotic cells and cross-presentation of antigen. Tim-3 functions to inhibit aggressive Th1-mediated auto- and alloimmune responses. Tim-3 pathway blockade by administration of Tim-3:Fc fusion protein accelerates diabetes in nonobese diabetic mice, causes hyperproliferation of Th1 cells and Th1 cytokine release in an experimental autoimmune encephalomyelitis (EAE) model and prevents acquisition of transplantation tolerance induced by costimulation blockade.