Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Tim-3 (from CHO Cells)
Human Recombinant Tim-3 (from CHO Cells)
Catalog # 102981-684
Supplier:  Adipogen
CAS Number:  
Human Recombinant Tim-3 (from CHO Cells)
Catalog # 102981-684
Supplier:  Adipogen
Supplier Number:  CHI-HF-211T3-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    <0.06EU/µg
  • Biological activity:
    Measured by its binding ability in a functional ELISA.
  • Gene ID:
    NP_116171.3
  • Protein synonyms:
    TIMD3|Hepatitis A Virus Cellular Receptor 2|HAVcr-2|T Cell Immunoglobulin and Mucin Domain-containing Protein 3|TIM3
  • Protein/peptide name:
    Tim-3
  • Purity:
    98%
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping temperature:
    -20 °C, Blue Ice
  • Cat. no.:
    102981-684
  • Supplier No.:
    CHIHF211T3C100

Specifications

About this item

The TIM (T cell/transmembrane, immunoglobulin and mucin) family plays a critical role in regulating immune responses, including allergy, asthma, transplant tolerance, autoimmunity and the response to viral infections. The unique structure of TIM immunoglobulin variable region domains allows highly specific recognition of phosphatidylserine (PtdSer), exposed on the surface of apoptotic cells. Tim-3, a type I transmembrane protein, contains an immunoglobulin and a mucin-like domain in its extracellular portion and a tyrosine phosphorylation motif in its cytoplasmic portion. TIM-3 is preferentially expressed on Th1 and Tc1 cells, and generates an inhibitory signal resulting in apoptosis of Th1 and Tc1 cells. TIM-3 is also expressed on some dendritic cells and can mediate phagocytosis of apoptotic cells and cross-presentation of antigen. Tim-3 functions to inhibit aggressive Th1-mediated auto- and alloimmune responses. Tim-3 pathway blockade by administration of Tim-3:Fc fusion protein accelerates diabetes in nonobese diabetic mice, causes hyperproliferation of Th1 cells and Th1 cytokine release in an experimental autoimmune encephalomyelitis (EAE) model and prevents acquisition of transplantation tolerance induced by costimulation blockade.