Human Recombinant IL-33R (from CHO Cells)
103691-928
: Adipogen
:
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Size:50 µg
- Storage conditions:–20 °C
- Endotoxin content:<0.06 EU/µg
- Biological activity:Measured by its ability to bind recombinant human IL-33 in a functional ELISA.
- Gene ID:AAH30975.1
- Protein synonyms:Interleukin-33 Receptor|ST2|DER4|IL1RL1
- Protein/peptide name:IL-33R
- Purity:98%
- Endotoxin level:Low
- Formulation:Lyophilized from 0.2 µm filtered solution in PBS.
- Shipping temperature:–20 °C, Blue Ice
- Cat. no.:103691-928
Receptor for interleukin-33 (IL-33). Its stimulation recruits MYD88, IRAK1, IRAK4 and TRAF6 followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14 and MAPK8 by similarity. Possibly involved in helper T cell function.