Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Size:50 µg
- Storage conditions:–20 °C
- Endotoxin content:<0.06EU/µg
- Gene ID:NP_663634.2
- Protein synonyms:Interleukin-27
- Protein/peptide name:IL-27
- Purity:98%
- Endotoxin level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping temperature:-20 °C, Blue Ice
- Cat. no.:102981-662
- Supplier No.:CHIHF21027C050
Specifications
About this item
Interleukin-27 (IL-27) is a heterodimeric group 2 receptor ligand molecule that belongs to the IL-6/IL-12 family of long type I cytokines. It is composed of EBI3 (EBV-induced gene 3), a 34 kDa glycoprotein that is related to the p40 subunit of IL-12 and IL-23, and p28, the cloned 28 kDa glycoprotein that is related to the p35 chain of IL-12. IL-27 is expressed by monocytes, endothelial cells and dendritic cells. IL-27 binds to and signals through a heterodimeric receptor complex composed of WSX1 (TCCR) and gp130. Evidence suggests IL-27 interacts only with WSX-1. IL-27 has both anti- and proinflammatory properties. As an antiinflammatory, IL-27 seems to induce a general negative feedback program that limits T and NK-T cell activity. At the onset of infection, IL-27 induces an IL-12 receptor on naïe CD4+ T cells, making them susceptible to subsequent IL-12 activity (and possible Th1 development). Notably, IL-12 family cytokines are both induced and inhibited by bacterial products. Microbes promote IL-27 secretion through TLR4, and also block IL-27 production via C5a induction.