Order Entry
Canada
ContactUsLinkComponent
Human Recombinant VEGF-C (from Insect Cells (Sf9))
Human Recombinant VEGF-C (from Insect Cells (Sf9))
Catalog # 102980-406
Supplier:  Adipogen
CAS Number:  
Human Recombinant VEGF-C (from Insect Cells (Sf9))
Catalog # 102980-406
Supplier:  Adipogen
Supplier Number:  AG-40T-0046-C020
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Size:
    20 µg
  • Storage conditions:
    –20 °C
  • Gene ID:
    P49767
  • Protein synonyms:
    Vascular Endothelial Growth Factor C|VRP|Flt4-L
  • Protein/peptide name:
    VEGF-C
  • Purity:
    90%
  • Molecular weight:
    ~18-24kDa
  • Formulation:
    Lyophilized
  • Shipping temperature:
    -20 °C, Blue Ice
  • Cat. no.:
    102980-406
  • Supplier No.:
    AG40T0046C020

Specifications

About this item

VEGF-C forms primarily non-covalently linked dimers. In adults, VEGF-C is highly expressed in heart, placenta, ovary and small intestine. It binds and activates VEGFR-2 (KDR/FLK1) and VEGFR-3 (FLT4) receptors. It is a growth factor active in angiogenesis and endothelial cell growth, stimulating their proliferation and migration and also has effects on the permeability of blood vessels. It is involved in the regulation of the growth and/or differentiation of lymphatic endothelium. It is also a mitogen for vascular endothelial cells.