About this item
Human hepatocyte growth factor (HGF) is synthesized as a biologically inactive single chain precursor, which is cleaved by a specific, extracellular serum serine protease to a fully active heterodimer. It stimulates the motility and invasion of several cancer cell types and can induce angiogenesis. All biological responses induced by HGF are elicited by binding to its transmembrane tyrosine kinase receptor. HGF function is essential for normal development. Knockout studies have demonstrated that both ligand and receptor deficient mice display an embryonic lethal phenotype. HGF synergizes with basic FGF in the induction of angiogenesis. HGF is a potent mitogen for mature parenchymal hepatocyte cells and is a pleiotrophic cytokine. It acts as growth factor for a broad spectrum of tissues and cell types. It has no detectable protease activity. It is an activating ligand for the receptor tyrosine kinase MET by binding and promoting its dimerization.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:–20 °C
- Biological activity:The activity was assayed for scattering activity in the MDCK cell assay. The ED50 for this effect is typically at 1.0-5.0ng/ml.
- Gene ID:P14210
- Protein synonyms:Hepatocyte Growth Factor|Scatter Factor|Hepatopoietin-A
- Protein/peptide name:HGF
- Purity:90%
- Molecular weight:~78kDa
- Formulation:Lyophilized
- Shipping temperature:-20 °C, Blue Ice