Order Entry
Canada
ContactUsLinkComponent
Human Recombinant NF-kappaB (active) (from Insect Cells (Sf21))
Human Recombinant NF-kappaB (active) (from Insect Cells (Sf21))
Catalog # 102980-376
Supplier:  Adipogen
CAS Number:  
Human Recombinant NF-kappaB (active) (from Insect Cells (Sf21))
Catalog # 102980-376
Supplier:  Adipogen
Supplier Number:  AG-40T-0020-C002
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Human
  • Size:
    2 µg
  • Storage conditions:
    –80 °C
  • Biological activity:
    ~1ng is required for high mobility shift assay.~5ng are required for reconstituted transcription assays.
  • Gene ID:
    Q04206
  • Protein synonyms:
    Transcription Factor p65|Nuclear Factor NF-kappaB p65 Subunit|RELA
  • Protein/peptide name:
    NF-kappaB (active)
  • Purity:
    95%
  • Concentration:
    Lot dependent (0.2-1.5mg/ml)
  • Formulation:
    Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 0.2% NP-40, 50mM imidazole and 10% glycerol.
  • Shipping temperature:
    -80 °C, Dry Ice
  • Cat. no.:
    102980-376
  • Supplier No.:
    AG40T0020C002

Specifications

About this item

NF-kappaB is a pleiotropic transcription factor present in almost all cell types and is the endpoint of a series of signal transduction events that are initiated by a vast array of stimuli related to many biological processes such as inflammation, immunity, differentiation, cell growth, tumorigenesis and apoptosis. NF-kappaB is a homo- or heterodimeric complex formed by the Rel-like domain-containing proteins RELA/p65, RELB, NFKB1/p105, NFKB1/p50, REL and NFKB2/p52. The heterodimeric p65-p50 complex is the most abundant complex. The dimers bind at kappaB sites in the DNA of their target genes and the individual dimers have distinct preferences for different kappaB sites that they can bind with distinguishable affinity and specificity.Different dimer combinations act as transcriptional activators or repressors, respectively. NF-kappaB complexes are held in the cytoplasm in an inactive state complexed with members of the NF-kappaB inhibitor (I-kappaB) family. In a conventional activation pathway, I-kappaB is phosphorylated by I-kappaB kinases (IKKs) in response to different activators, subsequently degraded thus liberating the active NF-kappaB complex which translocates to the nucleus. NF-kappaB heterodimeric p65-p50 and p65-c-Rel complexes are transcriptional activators. The NF-kappaB p65-p65 complex appears to be involved in invasin-mediated activation of IL-8 expression. p65 shows a weak DNA-binding site which could contribute directly to DNA binding in the NF-kappaB complex.