Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-33 (from E. coli)
Human Recombinant IL-33 (from E. coli)
Supplier:  Adipogen
Certificates
Human Recombinant IL-33 (from E. coli)
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Interleukins
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    <0.01EU/µg
  • Biological activity:
    Activates human ST2-dependent NF-kappaB pathway.
  • Gene ID:
    Q2YEJ5
  • Protein synonyms:
    Interleukin-33 (C208S/C232S Mutant)|IL-1F11|NF-HEV|IL-33 (C208S/C232S Mutant)
  • Protein/peptide name:
    IL-33
  • Purity:
    95%
  • Molecular weight:
    ~17kDa
  • Endotoxin level:
    Low
  • Concentration:
    After reconstitution:for 10µg size: 0.1mg/mlfor 100µg size: 1mg/ml
  • Formulation:
    Lyophilized. Contains PBS.
  • Shipping temperature:
    -20 °C, Blue Ice

Specifications

Frequently Bought Together