About this item
Collagen triple helix repeat containing-1 (CTHRC1) is a 30kDa glycosylated secretory protein containing a short collagen-like motif contributing to vascular remodeling by limiting collagen matrix deposition and promoting cell migration. CTHRC1 is expressed in calcified atherosclerotic plaque and chondrocyte-like cells, in the hypothalamus, pituitary gland and remodeling tissues. Aberrant expression of CTHRC1 is present in human solid cancers and likely to be associated with cancer invasion and metastasis. With adiponectin and other members of the C1q/TNF-related protein (CTRP) family, CTHRC1 shares the presence of a short N-terminal collagen domain but it lacks the characteristic C1q domain that defines the CTRP family of proteins. A recent study identifies CTHRC1 as a novel circulating hormone with metabolic effects.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:<0.01EU/µg
- Gene ID:Q96CG8
- Protein synonyms:NMTC1|Collagen Triple Helix Repeat-containing Protein 1
- Protein/peptide name:CTHRC1
- Purity:90%
- Molecular weight:~Two bands of 25-30 kDa (SDS-PAGE)
- Endotoxin level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping temperature:-20 °C, Blue Ice