About this item
Fibroblast growth factor 21 (FGF-21) is a member of the fibroblast growth factor family and has been identified as an important regulator of energy metabolism connecting nutrition, growth, reproduction and longevity. FGF-21 lacks the heparin-binding domain of most FGF proteins, allowing its secretion. FGF-21 is abundantly expressed in adipose tissues, liver and pancreas. In adipose tissue, FGF-21 promotes glucose uptake and oxidation and in liver it replenishes on fasting the tissues with fuel on low nutritional supply. It also induces lipolysis and ketogenesis. FGF-21 acts through FGF receptors (FGFR) associated with the auxiliary protein beta-Klotho. Recently, FGF-21 has been reported to interact directly with the brain circadian clock to coordinate activity and reproduction as part of the adaptation to fasting. Due to its multiple functions of normalizing glucose, lipid and energy homeostasis, FGF-21 represents an attractive novel therapy for type 2 diabetes mellitus and obesity.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:<0.01EU/µg
- Gene ID:Q9JJN1
- Protein synonyms:Fibroblast Growth Factor 21
- Protein/peptide name:FGF-21
- Purity:95%
- Molecular weight:~24kDa (SDS-PAGE)
- Endotoxin level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping temperature:-20 °C, Blue Ice