About this item
Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin). Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Storage conditions:–20 °C
- Endotoxin content:<0.01EU/µg
- Biological activity:Activates brown adipocyte markers Ucp1 and Cidec in murine subcutaneous adipocytes at 10ng/ml [Lit. 1].
- Gene ID:Q8NAU1
- Protein synonyms:FNDC5 (cleaved)|Fibronectin Type III Domain-containing Protein 5 (cleaved)|Fibronectin Type III Repeat-containing Protein 2 (cleaved)
- Protein/peptide name:Irisin
- Purity:95%
- Molecular weight:~26kDa
- Endotoxin level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping temperature:-20 °C, Blue Ice