About this item
Fc (human):FasL, Soluble (human) is a high activity construct in which two trimeric FasL are artificially linked via the Fc binding domain of human IgG1. This construct very effectively simulates the natural membrane-assisted aggregation of FasL in vivo. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. It is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:<0.01EU/µg
- Biological activity:Induces apoptosis of human Jurkat T cells at a concentration of <0.2ng/ml.
- Gene ID:P48023
- Protein synonyms:TNFSF6|CD178|APO-1L|CD95L|Fas Ligand
- Protein/peptide name:FasL (soluble)
- Purity:95%
- Molecular weight:~50kDa (SDS-PAGE)
- Endotoxin level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping temperature:-20 °C, Blue Ice