Order Entry
Canada
ContactUsLinkComponent
Human Recombinant FLT3 Ligand (from CHO cells)
Human Recombinant FLT3 Ligand (from CHO cells)
Supplier:  Adipogen
Certificates
Human Recombinant FLT3 Ligand (from CHO cells)
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    <0.1EU/µg
  • Biological activity:
    In vitro and in vivo generation of conventional and plasmacytoid dendritic cells (DC).
    FLT3L increases in vivo both NK and DC cells numbers in the spleen when mice are injected intraperitoneally with 2 or 10 µg, daily for 10 days (slightly higher effect at 10 µg).
  • Gene ID:
    P49771
  • Protein synonyms:
    FMS-like Tyrosine Kinase 3 Ligand
  • Protein/peptide name:
    FLT3 Ligand
  • Purity:
    95%
  • Endotoxin level:
    Low
  • Concentration:
    1mg/ml
  • Formulation:
    Liquid. In PBS (sterile).
  • Shipping temperature:
    -20 °C, Blue Ice

Specifications

Frequently Bought Together