About this item
IL-36Ra/IL-1F5 is a highly and a specific antagonist of the IL-1 receptor-related protein 2-mediated response to IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9). These cytokines bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1. IL-36Ra inhibits the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 induced by IL-36 in BMDC and CD4 T cells. Skin and dendritic cells are target of the IL-36 interleukins leading to a Th1 response. Recently mutations that affect the levels and the activity of IL-36Ra have been found in patients with pustular psoriasis, leading to enhanced production of inflammatory cytokines (IL-8 in particular) by keratinocytes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:<0.05EU/µg
- Gene ID:Q9UBH0
- Amino acid number:2 to 155
- Protein synonyms:Interleukin-1delta Interleukin-1 Homolog 3|IL-1delta|Interleukin-36 Receptor Antagonist Protein|Interleukin-1 Family Member 5|Interleukin-1-like Protein 1|IL-1F5
- Protein/peptide name:IL-36Ra
- Purity:95%
- Molecular weight:~17kDa
- Endotoxin level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping temperature:-20 °C, Blue Ice