Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Jagged-2 (from HEK293 cells)
Human Recombinant Jagged-2 (from HEK293 cells)
Catalog # 102980-082
Supplier:  Adipogen
CAS Number:  
Human Recombinant Jagged-2 (from HEK293 cells)
Catalog # 102980-082
Supplier:  Adipogen
Supplier Number:  AG-40A-0155Y-C010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    10 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    <0.1EU/µg
  • Gene ID:
    Q9Y219
  • Protein synonyms:
    hJ2|JAG2|Protein Jagged-2
  • Protein/peptide name:
    Jagged-2
  • Purity:
    90%
  • Molecular weight:
    ~150kDa (SDS-PAGE)
  • Endotoxin level:
    Low
  • Concentration:
    0.1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS + 0.02 % CHAPS.
  • Shipping temperature:
    -20 °C, Blue Ice
  • Cat. no.:
    102980-082
  • Supplier No.:
    AG40A0155YC010

Specifications

About this item

Jagged-2 is a putative Notch ligand involved in the mediation of Notch signaling, that can induce stromal cells to secrete IL-6, VEGF and IGF-1. Notch activation can interact with NF-kappaB and C-myc to promote the proliferation and to inhibit the apoptosis of MM cells, showing in the relationship between the incidence of myeloma and drug resistance. Jagged-2 is also involved in limb development.