About this item
DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:<0.1EU/µg
- Gene ID:Q8NFT8
- Protein synonyms:BET|Delta and Notch-like Epidermal Growth Factor-related Receptor|UNQ262/PRO299
- Protein/peptide name:DNER
- Purity:90%
- Molecular weight:~120-140kDa (SDS-PAGE)
- Endotoxin level:Low
- Concentration:0.1 mg /ml after reconstitution.
- Formulation:Lyophilized. Contains PBS + 0.02% CHAPS.
- Shipping temperature:-20 °C, Blue Ice