About this item
DLL1 (Delta-like protein; Delta1) is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK-cell precursor.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:<0.01EU/µg
- Biological activity:Inhibits adipogenesis of 3T3L-1 cells. Induces the Notch target gene HES-1 when coated on a plate at 1µg/ml.
- Gene ID:O00548
- Protein synonyms:Delta1|Delta-like Protein 1
- Protein/peptide name:DLL1
- Purity:90%
- Molecular weight:~100kDa (SDS-PAGE)
- Endotoxin level:Low
- Concentration:After reconstitution:for 10µg size: 0.1mg/mlfor 50µg size: 1 mg/ml
- Formulation:Lyophilized. Contains PBS.
- Shipping temperature:-20 °C, Blue Ice