Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen symbol:PEDF
- Clone:Serpy-1-4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293
- Isotype:IgG2 lambda
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:liquid
- Gene ID:P36955
- Antigen synonyms:Serpin F1|EPC-1|Pigment Epithelium-derived Factor|Cell Proliferation-inducing Gene 35 Protein
- Storage temperature:–20 °C
- Concentration:1mg/ml
- Shipping temperature:-20° C
- Immunogen:Human recombinant PEDF (AG-40B-0077).
- Cat. no.:102980-014
- Supplier No.:AG27B0014C100
Pigment epithelium-derived factor (PEDF) is a 47kDa secreted glycoprotein that belongs to the non-inhibitory serpin family group. PEDF is widely expressed in adult and fetal tissues, including brain, spinal cord, plasma, bone, prostate, pancreas, heart and lung. PEDF acts as an angiogenesis inhibitor with neurotrophic, immunomodulation and antitumor properties. It functions as anti-angiogenic agent by counterbalancing the proangiogenic effect of VEGF. PEDF is one of the most abundant proteins released by adipocytes and induces insulin resistance in adipocytes and human skeletal muscle cells. Recently, it has been reported that PEDF is sufficient to maintain the self-renewal of pluripotent human embryonic stem cells. Anti-PEDF (human), mAb (rec.) (Serpy-1-4) is an antibody developed by antibody phage display technology using a human naive antibody gene library. These libraries consist of scFv (single chain fragment variable) composed of VH (variable domain of the human immunoglobulin heavy chain) and VL (variable domain of the human immunoglobulin light chain) connected by a polypeptide linker. The antibody fragments are displayed on the surface of filamentous bacteriophage (M13). This scFv was selected by affinity selection on antigen in a process termed panning. Multiple rounds of panning are performed to enrich for antigen-specific scFv-phage. Monoclonal antibodies are subsequently identified by screening after each round of selection. The selected monoclonal scFv is cloned into an appropriate vector containing a Fc portion of interest and then produced in mammalian cells to generate an IgG like scFv-Fc fusion protein.
Type: Primary
Antigen: PEDF
Clonality: Recombinant
Clone: Serpy-1-4
Conjugation: Unconjugated
Epitope:
Host: Purified from HEK 293 cell culture supernatant.
Isotype: Human IgG2lambda
Reactivity: Human