MRO Plus® Heavyweight Universal Absorbent Roll, Brady® SPC® Absorbents
Catalog # 10810-844
Supplier: Brady Worldwide
Specifications
- Color:Gray
- Description:MRO Plus® Heavyweight Universal Absorbent Roll
- Length:150'
- Material:PP
- Cat. no.:10810-844
- Width:30"
- Supplier No.:MRO30P
Specifications
About this item
MRO Plus® products are strong, durable, and low linting, making them the perfect go-to products for nearly every industrial setting.
- Highly versatile, grey absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids
- Heavyweight design specifically targets continuous leaks and larger spills
- Three-ply meltblown polypropylene construction is designed for general industrial applications
- Perforations provide cost control and reduce waste by allowing users to only utilize amounts needed
The MRO Plus® Heavy Roll absorbs all industrial liquids (water, petroleum-based, and chemical-based fluids). Three-ply construction makes it stronger, more durable, and reduces lint. Perforated every 30".
Certifications: EPA 40 CFR 112.7(c)(1)(vii). OSHA 29 CFR 1910.120(j)(1)(vii). OSHA 29 CFR 1910.22(a)(2).