ALLWIK® Universal Absorbent Socks, Brady® SPC® Absorbents
10810-746
- Color:Gray
- Description:ALLWIK® universal absorbent socks
- Length:12'
- Material:PP
- Environmentally Preferable:
- Cat. no.:10810-746
- Supplier No.:AW124
- Diameter:3"
Use ALLWIK® all-purpose absorbents to clean up oils, coolants, solvents, and water-based fluids throughout any facility.
- Moldable SOCs wrap around machinery and conform to uneven surfaces to prevent leaks from reaching walkways
- Highly versatile, grey absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids
- Gray color masks spills and stains, helping users resist the urge to pick up the absorbent before it is fully saturated
- Made of all-purpose polypropylene, suitable for most industrial applications
Polypropylene ALLWIK® SOCs are wringable, incinerable, and suitable for most industrial applications.
: EPA 40 CFR 112.7(c)(1)(vii). OSHA 29 CFR 1910.120(j)(1)(vii). OSHA 29 CFR 1910.22(a)(2).